siRNA Details
| Gene Symbol: Gm5111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183309.3 | NP_899132.1 | 2 | 387 | ||
| GenBank mRNA | AK041789.1 | - | 2 | 387 | ||
| BC104366.1 | - | 2 | 382 | |||
| BC104367.1 | - | 1 | 416 | |||
| BC127138.1 | - | 1 | 418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5111| Gene Symbol: Gm5111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183309.3 | NP_899132.1 | 2 | 387 | ||
| GenBank mRNA | AK041789.1 | - | 2 | 387 | ||
| BC104366.1 | - | 2 | 382 | |||
| BC104367.1 | - | 1 | 416 | |||
| BC127138.1 | - | 1 | 418 | |||