siRNA Details
| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142781.1 | NP_001136253.1 | 4 | 581 | ||
| NM_177883.4 | NP_808551.2 | 4 | 581 | |||
| GenBank mRNA | AK082855.1 | - | 4 | 582 | ||
| AK142385.1 | - | 4 | 499 | |||
| AK161304.1 | - | 4 | 459 | |||
| BC118939.1 | - | 4 | 575 | |||
| BC118940.1 | - | 4 | 575 | 40092089 | ||

Fam188b
Genomic Map