siRNA Details
| Gene Symbol: Gcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177884.3 | NP_808552.2 | 14, 15 | 1959 | ||
| GenBank mRNA | AK038369.1 | - | 15, 16 | 1946 | ||
| AK050789.1 | - | 14, 15 | 1901 | |||
| AK078171.1 | - | 14, 15 | 1949 | |||
| AK136387.1 | - | 12, 13 | 1561 | |||

Gcfc2
Genomic Map