siRNA Details
| Gene Symbol: Fbxo41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001160.3 | NP_001001160.1 | 8 | 2441 | ||
| NM_001289674.1 | NP_001276603.1 | 7 | 2272 | |||
| NM_001289675.1 | NP_001276604.1 | 7 | 2266 | |||
| GenBank mRNA | AK129466.1 | - | 6 | 1818 | ||
| AK147230.1 | - | 8 | 2333 | |||
| AK147681.1 | - | 7 | 2131 | |||
| BC061475.1 | - | 5 | 913 | |||
| BC070445.1 | - | 7 | 2272 | 30536738 | ||

Fbxo41
Genomic Map