siRNA Details
| Gene Symbol: Tmcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177412.1 | NP_803131.1 | 3 | 660 | ||
| GenBank mRNA | AK087243.1 | - | 1 | 1153 | ||
| AK163297.1 | - | 3 | 596 | |||
| AK173022.1 | - | 3 | 649 | |||
| BC029759.1 | - | 1 | 350 | |||
| BC157922.1 | - | 1 | 460 | |||

Tmcc1
Genomic Map