siRNA Details
| Gene Symbol: Tmcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177412.1 | NP_803131.1 | 4 | 1448 | ||
| GenBank mRNA | AK028322.1 | - | 2 | 1077 | ||
| AK154540.1 | - | 2 | 1134 | |||
| AK173022.1 | - | 4 | 1437 | |||
| BC029759.1 | - | 3 | 1270 | |||
| BC157922.1 | - | 3 | 1371 | |||

Tmcc1
Genomic Map