siRNA Details
| Gene Symbol: B4galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198884.1 | NP_942585.1 | 5, 6 | 533 | ||
| GenBank mRNA | AB114826.1 | - | 5, 6 | 533 | ||
| AK143898.1 | - | 5, 6 | 447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B4galnt3| Gene Symbol: B4galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198884.1 | NP_942585.1 | 5, 6 | 533 | ||
| GenBank mRNA | AB114826.1 | - | 5, 6 | 533 | ||
| AK143898.1 | - | 5, 6 | 447 | |||