siRNA Details
| Gene Symbol: B4galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198884.1 | NP_942585.1 | 14 | 1828 | ||
| GenBank mRNA | AB114826.1 | - | 14 | 1828 | ||
| AK143898.1 | - | 14 | 1744 | |||
| BC139798.1 | - | 14 | 1842 | 40060840 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B4galnt3| Gene Symbol: B4galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198884.1 | NP_942585.1 | 14 | 1828 | ||
| GenBank mRNA | AB114826.1 | - | 14 | 1828 | ||
| AK143898.1 | - | 14 | 1744 | |||
| BC139798.1 | - | 14 | 1842 | 40060840 | ||