siRNA Details
| Gene Symbol: Tmem52b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081186.1 | NP_001074655.1 | 4, 5 | 790 | ||
| GenBank mRNA | AK085389.1 | - | 4, 5 | 794 | ||
| AK085571.1 | - | 4, 5 | 731 | |||
| BC120661.1 | - | 4, 5 | 443 | 40129584 | ||
| BC120663.1 | - | 4, 5 | 443 | 40129586 | ||
| BC144939.1 | - | 3, 4 | 399 | |||

Tmem52b
Genomic Map