siRNA Details
| Gene Symbol: RAB3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002866.4 | NP_002857.1 | 3 | 434 | ||
| XM_011528164.1 | XP_011526466.1 | 3 | 431 | |||
| GenBank mRNA | AF254795.1 | - | 3 | 394 | ||
| AF498931.1 | - | 2 | 265 | |||
| AK289559.1 | - | 3 | 412 | |||
| AK293467.1 | - | 408 | ||||
| BC011782.2 | - | 3 | 399 | 3939176 | ||
| DB497194.1 | - | 3 | 444 | |||
| M28210.1 | - | 3 | 304 | |||
| X13900.1 | - | 3 | 304 | |||

RAB3A
Genomic Map