siRNA Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198631.2 | NP_941033.2 | 4 | 530 | ||
| GenBank mRNA | AK151148.1 | - | 5 | 582 | ||
| AK151306.1 | - | 5 | 582 | |||
| BC057627.1 | - | 4 | 475 | 6412890 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h4| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198631.2 | NP_941033.2 | 4 | 530 | ||
| GenBank mRNA | AK151148.1 | - | 5 | 582 | ||
| AK151306.1 | - | 5 | 582 | |||
| BC057627.1 | - | 4 | 475 | 6412890 | ||