siRNA Details
Gene Symbol: Tmem145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183311.2 | NP_899134.2 | 13 | 1242 | ||
GenBank mRNA | AK081054.1 | - | 13 | 1243 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183311.2 | NP_899134.2 | 13 | 1242 | ||
GenBank mRNA | AK081054.1 | - | 13 | 1243 |