siRNA Details
| Gene Symbol: Ifnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024673.2 | NP_001019844.2 | 5 | 523 | ||
| GenBank mRNA | AY869695.1 | - | 5 | 523 | ||
| BC150796.1 | - | 5 | 618 | 9087707 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifnl2| Gene Symbol: Ifnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024673.2 | NP_001019844.2 | 5 | 523 | ||
| GenBank mRNA | AY869695.1 | - | 5 | 523 | ||
| BC150796.1 | - | 5 | 618 | 9087707 | ||