siRNA Details
| Gene Symbol: Zfp82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252519.1 | NP_001239448.1 | 6 | 1758 | ||
| NM_177889.5 | NP_808557.2 | 7 | 1848 | |||
| GenBank mRNA | AK129467.1 | - | 2 | 3259 | ||
| AK137630.1 | - | 1472 | ||||
| BC025629.1 | - | 1 | 1004 | |||
| BC060530.1 | - | 6 | 1775 | 5684819 | ||
| BX525285.1 | - | 1 | 151 | |||

Zfp82
Genomic Map