siRNA Details
| Gene Symbol: Fan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177893.3 | NP_808561.2 | 2 | 578 | ||
| GenBank mRNA | AK077918.1 | - | 2 | 569 | ||
| AK173087.1 | - | 1 | 36 | |||
| BC116270.2 | - | 1 | 704 | |||
| BC116271.2 | - | 1 | 704 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fan1| Gene Symbol: Fan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177893.3 | NP_808561.2 | 2 | 578 | ||
| GenBank mRNA | AK077918.1 | - | 2 | 569 | ||
| AK173087.1 | - | 1 | 36 | |||
| BC116270.2 | - | 1 | 704 | |||
| BC116271.2 | - | 1 | 704 | |||