siRNA Details
| Gene Symbol: Saxo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177894.4 | NP_808562.3 | 2, 3 | 351 | ||
| GenBank mRNA | AK051095.1 | - | 2, 3 | 347 | ||
| BC144779.1 | - | 2, 3 | 296 | |||
| BC144780.1 | - | 2, 3 | 313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Saxo2| Gene Symbol: Saxo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177894.4 | NP_808562.3 | 2, 3 | 351 | ||
| GenBank mRNA | AK051095.1 | - | 2, 3 | 347 | ||
| BC144779.1 | - | 2, 3 | 296 | |||
| BC144780.1 | - | 2, 3 | 313 | |||