siRNA Details
| Gene Symbol: Saxo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177894.4 | NP_808562.3 | 2 | 238 | ||
| GenBank mRNA | AK038727.1 | - | 3 | 418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Saxo2| Gene Symbol: Saxo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177894.4 | NP_808562.3 | 2 | 238 | ||
| GenBank mRNA | AK038727.1 | - | 3 | 418 | ||