siRNA Details
| Gene Symbol: Nek5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177898.4 | NP_808566.2 | 2 | 157 | ||
| GenBank mRNA | AK032672.1 | - | 2 | 159 | ||
| AK054168.1 | - | 2 | 171 | |||
| BC053516.1 | - | 2 | 145 | 6311513 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nek5| Gene Symbol: Nek5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177898.4 | NP_808566.2 | 2 | 157 | ||
| GenBank mRNA | AK032672.1 | - | 2 | 159 | ||
| AK054168.1 | - | 2 | 171 | |||
| BC053516.1 | - | 2 | 145 | 6311513 | ||