siRNA Details
| Gene Symbol: Rnf150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177378.4 | NP_796352.2 | 2 | 1341 | ||
| GenBank mRNA | AK041412.1 | - | 2 | 1343 | ||
| AK220374.1 | - | 2 | 1224 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf150| Gene Symbol: Rnf150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177378.4 | NP_796352.2 | 2 | 1341 | ||
| GenBank mRNA | AK041412.1 | - | 2 | 1343 | ||
| AK220374.1 | - | 2 | 1224 | |||