siRNA Details
| Gene Symbol: Adgrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181039.2 | NP_851382.2 | 5 | 1722 | ||
| GenBank mRNA | AK147455.1 | - | 5 | 1716 | ||
| AK147519.1 | - | 3 | 629 | |||
| BC085138.1 | - | 6 | 1123 | |||
More Information
Set Membership: |
|

Adgrl1
Genomic Map