siRNA Details
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042715.3 | NP_001036180.2 | 14 | 2060 | ||
| GenBank mRNA | AY352586.2 | - | 14 | 2057 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Drc7| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042715.3 | NP_001036180.2 | 14 | 2060 | ||
| GenBank mRNA | AY352586.2 | - | 14 | 2057 | ||