siRNA Details
| Gene Symbol: Arhgap32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195632.1 | NP_001182561.1 | 12 | 1207 | ||
| NM_177379.4 | NP_796353.3 | 3 | 543 | |||
| GenBank mRNA | AB088417.1 | - | 4 | 490 | ||
| AK036860.1 | - | 1 | 42 | |||
| AY194286.1 | - | 3 | 270 | |||
| BC132390.1 | - | 2 | 199 | 40130667 | ||
| BC138042.1 | - | 2 | 199 | 8861061 | ||

Arhgap32
Genomic Map