siRNA Details
| Gene Symbol: AI593442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286641.1 | NP_001273570.1 | 2 | 2017 | ||
| NM_178906.5 | NP_849237.2 | 2 | 1968 | |||
| GenBank mRNA | AK033979.1 | - | 2 | 1962 | ||
| AK053489.1 | - | 1 | 104 | |||
| CO039428.1 | - | 1 | 122 | |||

AI593442
Genomic Map