siRNA Details
| Gene Symbol: Tgm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177911.4 | NP_808579.2 | 11 | 1442 | ||
| GenBank mRNA | AF486627.1 | - | 11 | 1441 | ||
| AK035279.1 | - | 11 | 1443 | |||
| BC141297.1 | - | 11 | 1439 | 9055909 | ||
| BC145622.1 | - | 11 | 1439 | 9054190 | ||

Tgm4
Genomic Map