siRNA Details
Gene Symbol: Thoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033422.1 | NP_001028594.1 | 14, 15 | 1592 | ||
GenBank mRNA | AK140950.1 | - | 14, 15 | 1584 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Thoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033422.1 | NP_001028594.1 | 14, 15 | 1592 | ||
GenBank mRNA | AK140950.1 | - | 14, 15 | 1584 |