siRNA Details
Gene Symbol: Thoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033422.1 | NP_001028594.1 | 11 | 1193 | ||
GenBank mRNA | AK138023.1 | - | 11 | 1194 | ||
AK140950.1 | - | 11 | 1185 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Thoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033422.1 | NP_001028594.1 | 11 | 1193 | ||
GenBank mRNA | AK138023.1 | - | 11 | 1194 | ||
AK140950.1 | - | 11 | 1185 |