siRNA Details
| Gene Symbol: Gm5127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033541.2 | NP_001028713.1 | 4 | 794 | ||
| GenBank mRNA | AK044840.1 | - | 7 | 1136 | ||
| AK142438.1 | - | 4 | 795 | |||
| BC147417.1 | - | 1 | 469 | 9056484 | ||
| BC147418.1 | - | 1 | 469 | 9056504 | ||

Gm5127
Genomic Map