siRNA Details
Gene Symbol: Gm5128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183320.3 | NP_899143.2 | 4 | 611 | ||
GenBank mRNA | AK029697.1 | - | 612 | |||
BC115710.1 | - | 606 | 40058794 |
Gene Symbol: Gm7903 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242937.1 | NP_001229866.1 | 611 |
