siRNA Details
| Gene Symbol: AV320801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177918.1 | NP_808586.1 | 540 | |||
| GenBank mRNA | AK029939.1 | - | 541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AV320801| Gene Symbol: AV320801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177918.1 | NP_808586.1 | 540 | |||
| GenBank mRNA | AK029939.1 | - | 541 | |||