siRNA Details
| Gene Symbol: Tceal5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177919.2 | NP_808587.1 | 3 | 930 | ||
| GenBank mRNA | AK032140.1 | - | 3 | 932 | ||
| BC100415.1 | - | 3 | 955 | 6535676 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tceal5| Gene Symbol: Tceal5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177919.2 | NP_808587.1 | 3 | 930 | ||
| GenBank mRNA | AK032140.1 | - | 3 | 932 | ||
| BC100415.1 | - | 3 | 955 | 6535676 | ||