siRNA Details
| Gene Symbol: Tceal5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177919.2 | NP_808587.1 | 3 | 244 | ||
| GenBank mRNA | AK032140.1 | - | 3 | 246 | ||
| BC100415.1 | - | 3 | 269 | 6535676 | ||
| BC115460.1 | - | 2 | 155 | 40100235 | ||
| BC115461.1 | - | 2 | 155 | 40100237 | ||

Tceal5
Genomic Map