siRNA Details
| Gene Symbol: E230019M04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177921.3 | NP_808589.1 | 2, 3 | 259 | ||
| GenBank mRNA | AK054107.1 | - | 2, 3 | 260 | ||
| BC119078.1 | - | 2, 3 | 256 | 8733827 | ||
| BC120495.1 | - | 2, 3 | 256 | 40129418 | ||

E230019M04Rik
Genomic Map