siRNA Details
| Gene Symbol: Zdhhc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007460.1 | NP_001007461.1 | 1 | 59 | ||
| GenBank mRNA | AK147553.1 | - | 2 | 420 | ||
| AY668949.1 | - | 1 | 59 | |||
| BC139052.1 | - | 1 | 102 | 8862074 | ||
| BC145525.1 | - | 1 | 102 | 9054088 | ||
| BC145526.1 | - | 1 | 102 | 9054089 | ||
| BC145527.1 | - | 1 | 102 | |||
| BC145684.1 | - | 1 | 102 | 40130987 | ||

Zdhhc23
Genomic Map