siRNA Details
| Gene Symbol: Zscan10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033425.4 | NP_001028597.2 | 6 | 1104 | ||
| NM_001289481.1 | NP_001276410.1 | 6 | 1104 | |||
| NM_001289482.1 | NP_001276411.1 | 4 | 774 | |||
| NM_001289483.1 | NP_001276412.1 | 6 | 600 | |||
| NM_001289484.1 | NP_001276413.1 | 6 | 969 | |||
| GenBank mRNA | AK141259.1 | - | 6 | 1075 | ||
| BC068284.1 | - | 5 | 777 | |||
| BC145638.1 | - | 5 | 937 | 9054214 | ||
| DQ323929.1 | - | 6 | 1072 | |||
| DQ323930.1 | - | 6 | 1072 | |||
| DQ323931.1 | - | 4 | 742 | |||
| EF152498.1 | - | 5 | 919 | |||
| EF152499.1 | - | 6 | 564 | |||
| EF152500.1 | - | 6 | 933 | |||
| EF152501.1 | - | 4 | 696 | |||
| EF152502.1 | - | 4 | 696 | |||
| EF152503.1 | - | 2 | 573 | |||
| EF152504.1 | - | 3 | 630 | |||

Zscan10
Genomic Map