siRNA Details
| Gene Symbol: Grxcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033426.2 | NP_001028598.1 | 2 | 432 | ||
| GenBank mRNA | AK158418.1 | - | 2 | 433 | ||
| AK158432.1 | - | 317 | ||||
| BC147102.1 | - | 2 | 427 | 9056155 | ||
| BC147103.1 | - | 2 | 427 | 9056135 | ||

Grxcr2
Genomic Map