siRNA Details
| Gene Symbol: Grxcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033426.2 | NP_001028598.1 | 1 | 306 | ||
| GenBank mRNA | AK158418.1 | - | 1 | 307 | ||
| AK158432.1 | - | 191 | ||||
| BC147102.1 | - | 1 | 301 | 9056155 | ||
| BC147103.1 | - | 1 | 301 | 9056135 | ||

Grxcr2
Genomic Map