siRNA Details
Gene Symbol: BC051628 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199312.3 | NP_955016.1 | 2 | 1009 | ||
GenBank mRNA | BC051628.1 | - | 2 | 1009 | 6742098 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC051628 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199312.3 | NP_955016.1 | 2 | 1009 | ||
GenBank mRNA | BC051628.1 | - | 2 | 1009 | 6742098 |