siRNA Details
| Gene Symbol: Gm853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034872.2 | NP_001030044.1 | 7 | 1017 | ||
| GenBank mRNA | AK143920.1 | - | 7 | 1018 | ||
| BC147394.1 | - | 6 | 882 | 9056469 | ||
| BC147395.1 | - | 6 | 882 | 9056458 | ||
| BC172060.1 | - | 6 | 882 | 9054346 | ||

Gm853
Genomic Map