siRNA Details
| Gene Symbol: Tmem211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033428.2 | NP_001028600.1 | 3 | 393 | ||
| GenBank mRNA | AK138479.1 | - | 3 | 394 | ||
| AK138629.1 | - | 3 | 394 | |||
| BC147409.1 | - | 2 | 256 | 9056497 | ||
| BC147410.1 | - | 2 | 256 | 9056496 | ||
| BC172062.1 | - | 2 | 256 | 9054348 | ||

Tmem211
Genomic Map