siRNA Details
| Gene Symbol: Ksr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114545.2 | NP_001108017.2 | 2 | 1125 | ||
| GenBank mRNA | AK137433.1 | - | 2 | 1127 | ||
| BC150883.1 | - | 2 | 1125 | |||
| BC150890.1 | - | 2 | 1125 | |||
| DQ531035.1 | - | 2 | 298 | |||

Ksr2
Genomic Map