siRNA Details
| Gene Symbol: Kcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029985.4 | NP_001025156.3 | 18 | 1888 | ||
| GenBank mRNA | AK154369.1 | - | 18 | 1876 | ||
| AY884211.1 | - | 18 | 1874 | |||
| BQ179923.1 | - | 1 | 59 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcp| Gene Symbol: Kcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029985.4 | NP_001025156.3 | 18 | 1888 | ||
| GenBank mRNA | AK154369.1 | - | 18 | 1876 | ||
| AY884211.1 | - | 18 | 1874 | |||
| BQ179923.1 | - | 1 | 59 | |||