siRNA Details
| Gene Symbol: Cox6b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289848.1 | NP_001276777.1 | 2 | 257 | ||
| NM_001289849.1 | NP_001276778.1 | 2 | 226 | |||
| NM_001289850.1 | NP_001276779.1 | 2 | 226 | |||
| NM_183405.2 | NP_899664.1 | 2 | 226 | |||
| NM_183406.3 | NP_899665.1 | 1 | 137 | |||
| GenBank mRNA | AK160152.1 | - | 2 | 156 | ||
| AY152400.1 | - | 2 | 226 | |||
| BC048670.1 | - | 1 | 137 | 6773974 | ||
| BC107296.1 | - | 2 | 183 | 40053107 | ||
| BC107297.1 | - | 2 | 183 | 40053112 | ||
| BC125562.1 | - | 2 | 166 | 40130077 | ||
| BC125564.1 | - | 2 | 169 | 40130079 | ||
| BC145201.1 | - | 2 | 201 | 9053752 | ||
| BG080371.2 | - | 2 | 134 | |||
| CT010366.1 | - | 1 | 70 | |||
| W88135.1 | - | 1 | 214 | |||

Cox6b2
Genomic Map