siRNA Details
Gene Symbol: Proser3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183321.1 | NP_899144.1 | 10 | 1812 | ||
GenBank mRNA | BC053749.1 | - | 10 | 1812 | 6331972 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Proser3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183321.1 | NP_899144.1 | 10 | 1812 | ||
GenBank mRNA | BC053749.1 | - | 10 | 1812 | 6331972 |