siRNA Details
Gene Symbol: Trim75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033429.2 | NP_001028601.1 | 2 | 1299 | ||
GenBank mRNA | AK136018.1 | - | 2 | 1301 | ||
AK162165.1 | - | 1 | 142 | |||
AK163251.1 | - | 1 | 656 | |||
BC139219.1 | - | 1 | 1006 | 8862241 | ||
BC139221.1 | - | 1 | 1006 | 8862243 |
