siRNA Details
| Gene Symbol: Zfp36l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009549.2 | NP_001009549.1 | 1 | 2251 | ||
| GenBank mRNA | AV223369.2 | - | 1 | 245 | ||
| AY661338.1 | - | 1 | 2251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp36l3| Gene Symbol: Zfp36l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009549.2 | NP_001009549.1 | 1 | 2251 | ||
| GenBank mRNA | AV223369.2 | - | 1 | 245 | ||
| AY661338.1 | - | 1 | 2251 | |||