siRNA Details
| Gene Symbol: 4930453H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252013.1 | NP_001238942.1 | 5 | 1047 | ||
| GenBank mRNA | AK015451.1 | - | 5 | 1049 | ||
| Gene Symbol: Gm15104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101501.1 | NP_001094971.1 | 1 | 63 | ||
| GenBank mRNA | BC099570.1 | - | 2 | 140 | ||

4930453H23Rik
Genomic Map