siRNA Details
| Gene Symbol: Frmpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033330.3 | NP_001028502.1 | 13 | 1696 | ||
| NM_001290427.1 | NP_001277356.1 | 11 | 1307 | |||
| NM_001290428.1 | NP_001277357.1 | 10 | 1240 | |||
| GenBank mRNA | AK147370.1 | - | 13 | 1697 | ||
| AK147440.1 | - | 10 | 1241 | |||
| BC139429.1 | - | 11 | 1152 | 8862451 | ||

Frmpd4
Genomic Map