siRNA Details
| Gene Symbol: Frmpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033330.3 | NP_001028502.1 | 18 | 2478 | ||
| NM_001290427.1 | NP_001277356.1 | 16 | 2089 | |||
| NM_001290428.1 | NP_001277357.1 | 15 | 2022 | |||
| GenBank mRNA | AK122244.1 | - | 2 | 256 | ||
| AK147370.1 | - | 18 | 2479 | |||
| AK147440.1 | - | 15 | 2023 | |||
| BC139429.1 | - | 16 | 1934 | 8862451 | ||

Frmpd4
Genomic Map