siRNA Details
| Gene Symbol: Frmpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033330.3 | NP_001028502.1 | 18 | 2443 | ||
| NM_001290427.1 | NP_001277356.1 | 16 | 2054 | |||
| NM_001290428.1 | NP_001277357.1 | 15 | 1987 | |||
| GenBank mRNA | AK122244.1 | - | 2 | 221 | ||
| AK147370.1 | - | 18 | 2444 | |||
| AK147440.1 | - | 15 | 1988 | |||
| BC139429.1 | - | 16 | 1899 | 8862451 | ||

Frmpd4
Genomic Map