siRNA Details
| Gene Symbol: Ttc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290563.1 | NP_001277492.1 | 12, 13 | 1905 | ||
| NM_001290564.1 | NP_001277493.1 | 12, 13 | 1960 | |||
| NM_001290566.1 | NP_001277495.1 | 11, 12 | 1658 | |||
| NM_001290567.1 | NP_001277496.1 | 12, 13 | 1706 | |||
| NM_177384.3 | NP_796358.2 | 12, 13 | 1905 | |||
| GenBank mRNA | AK028040.1 | - | 12, 13 | 1960 | ||
| AK089384.1 | - | 9, 10 | 2761 | |||
| AK133273.1 | - | 12, 13 | 1683 | |||
| AK160435.1 | - | 12, 13 | 1656 | |||

Ttc16
Genomic Map